MilliporeSigma
  • Solution and mechanochemical syntheses, and spectroscopic and structural studies in the silver(I) (bi-)carbonate: triphenylphosphine system.
Solution and mechanochemical syntheses, and spectroscopic and structural studies in the silver(I) (bi-)carbonate: triphenylphosphine system.
Dalton transactions (Cambridge, England : 2003) (2011-06-11)
Graham A Bowmaker, Effendy, John V Hanna, Peter C Healy, Scott P King, Claudio Pettinari, Brian W Skelton, Allan H White
ABSTRACT
Syntheses of a number of adducts of silver(I) (bi-)carbonate with triphenylphosphine, both mechanochemically, and from solution, are described, together with their infra-red spectra, (31)P CP MAS NMR and crystal structures. Ag(HCO(3)):PPh(3) (1:4) has been isolated in the ionic form [Ag(PPh(3))(4)](HCO(3))·2EtOH·3H(2)O. Ag(2)CO(3):PPh(3) (1:4) forms a binuclear neutral molecule [(Ph(3)P)(2)Ag(O,μ-O'·CO)Ag(PPh(3))(2)](·2H(2)O), while Ag(HCO(3)):PPh(3) (1:2) has been isolated in both mononuclear and binuclear forms: [(Ph(3)P)(2)Ag(O(2)COH)] and [(Ph(3)P)(2)Ag(μ-O·CO·OH)(2)Ag(PPh(3))(2)] (both unsolvated). A more convenient method for the preparation of the previously reported copper(I) complex [(Ph(3)P)(2)Cu(HCO(3))] is also reported.
Sigma-Aldrich
Triphenylphosphine, ≥95.0% (GC)
Sigma-Aldrich
Triphenylphosphine, ReagentPlus®, 99%