MilliporeSigma
  • Acetylcholinesterase biosensor based on single-walled carbon nanotubes--Co phtalocyanine for organophosphorus pesticides detection.
Acetylcholinesterase biosensor based on single-walled carbon nanotubes--Co phtalocyanine for organophosphorus pesticides detection.
Talanta (2011-06-08)
A N Ivanov, R R Younusov, G A Evtugyn, F Arduini, D Moscone, G Palleschi
ABSTRACT
A simple and reliable technique has been developed for the construction of an amperometric acetylcholinesterase biosensor based on screen-printed carbon electrodes. For the first time, one-step modification using single-walled carbon nanotubes and Co phtalocyanine has been proposed to decrease the working potential and to increase the signal of thiocholine oxidation. The biosensor developed made it possible to detect 5-50 ppb of paraoxon and 2-50 ppb of malaoxon with detection limits of 3 and 2 ppb, respectively (incubation 15 min). The biosensor showed high reproducibility when measurements of the substrate and inhibitor were performed (R.S.D. about 1% and 2.5%, respectively). The reliability of the inhibition measurements was confirmed by testing spiked samples of sparkling and tape waters.
Product Description
Supelco
Malaoxon, PESTANAL®, analytical standard
Malathion impurity B, European Pharmacopoeia (EP) Reference Standard