MilliporeSigma
  • Chemical diversity of Ziziphora clinopodioides: composition of the essential oil of Z. clinopodioides from Tajikistan.
Chemical diversity of Ziziphora clinopodioides: composition of the essential oil of Z. clinopodioides from Tajikistan.
Natural product communications (2011-05-28)
Farukh S Sharopov, William N Setzer
ABSTRACT
The chemical composition of the essential oils of Ziziphora clinopodioides Lam. from the aerial flowering parts, collected during two different years, were obtained by hydrodistillation and analyzed by gas chromatography - mass spectrometry. Forty-five components representing 100% and 94.7% of the total oil were identified. The main constituents of the essential oils were pulegone (72.8 and 35.0%), neomenthol (6.5 and 23.1%), menthone (6.2 and 13.3%), p-menth-3-en-8-ol (1.7 and 3.5%), piperitenone (2.6 and 1.1%) and piperitone (0.7 and 1.2%). A cluster analysis was carried out on the essential oil compositions of Z. clinopodioides.
Product Description
Supelco
(+)-Pulegone, analytical standard
Sigma-Aldrich
(R)-(+)-Pulegone, ≥90%
Sigma-Aldrich
(R)-(+)-Pulegone, 85%, technical grade