MilliporeSigma
Triphenylene-fused porphyrins.
Organic letters (2011-05-25)
Lin Jiang, James T Engle, Laura Sirk, C Scott Hartley, Christopher J Ziegler, Hong Wang
ABSTRACT
Triphenylene has been successfully fused to the porphyrin periphery through a convenient oxidative ring-closure reaction. Bistriphenylene-fused porphyrins and a dibenzo[fg,op]tetracene-fused porphyrin have also been obtained using a similar approach. These π-extended porphyrins exhibited broadened and bathochromic shifted UV-vis absorptions.
Sigma-Aldrich
Triphenylene, 98%
Triphenylene, BCR®, certified reference material