MilliporeSigma
  • Hepatoprotective effects of polyprenols from Ginkgo biloba L. leaves on CCl4-induced hepatotoxicity in rats.
Hepatoprotective effects of polyprenols from Ginkgo biloba L. leaves on CCl4-induced hepatotoxicity in rats.
Fitoterapia (2011-05-21)
Lan Yang, Cheng-zhang Wang, Jian-zhong Ye, Hai-tao Li
ABSTRACT
The hepatoprotective effects of polyprenols from Ginkgo biloba L. leaves were evaluated against carbon tetrachloride induced hepatic damage in Sprague-Dawley rats. The elevated levels of serum ALT, AST, ALP, ALB, TP, HA, LN, TG, and CHO were restored towards normalization significantly by GBP in a dose dependent manner. The biochemical observations were supplemented with histopathological examination of rat liver sections. Meanwhile, GBP also produced a significant and dose-dependent reversal of CCl(4)-diminished activity of the antioxidant enzymes and reduced CCl(4)-elevated level of MDA. In general, the effects of GBP were not significantly different from those of the standard drug Essentiale.
Sigma-Aldrich
3-Methyl-2-buten-1-ol, 99%
Sigma-Aldrich
3-Methyl-2-buten-1-ol, ≥98%, FG