MilliporeSigma
  • p-hydroxyacetophenone amides from Cystobacter ferrugineus, strain Cb G35.
p-hydroxyacetophenone amides from Cystobacter ferrugineus, strain Cb G35.
Journal of natural products (2011-05-20)
Wiebke Zander, Kathrin I Mohr, Klaus Gerth, Rolf Jansen, Rolf Müller
ABSTRACT
A family of six novel p-hydroxyacetophenone amides, 1-6, was isolated from Cystobacter ferrugineus, strain Cb G35. Their structures were elucidated by ESI-TOF mass spectrometry and NMR spectroscopy. Feeding experiments with labeled [¹³C₉,¹⁵N]-tyrosine and [d₁₀]-leucine identified the biosynthetic precursors of 1.
Sigma-Aldrich
4′-Hydroxyacetophenone, 99%