MilliporeSigma
  • Biological and physicochemical properties of carbon-graphite fibre-reinforced polymers intended for implant suprastructures.
Biological and physicochemical properties of carbon-graphite fibre-reinforced polymers intended for implant suprastructures.
European journal of oral sciences (2011-05-14)
Susanna Segerström, Gunilla Sandborgh-Englund, Eystein I Ruyter
ABSTRACT
The aim of this study was to determine water sorption, water solubility, dimensional change caused by water storage, residual monomers, and possible cytotoxic effects of heat-polymerized carbon-graphite fibre-reinforced composites with different fibre loadings based on methyl methacrylate/poly(methyl methacrylate) (MMA/PMMA) and the copolymer poly (vinyl chloride-co-vinyl acetate). Two different resin systems were used. Resin A contained ethylene glycol dimethacrylate (EGDMA) and 1,4-butanediol dimethacrylate (1,4-BDMA); the cross-linker in Resin B was diethylene glycol dimethacrylate (DEGDMA). The resin mixtures were reinforced with 24, 36 and 47 wt% surface-treated carbon-graphite fibres. In addition, polymer B was reinforced with 58 wt% fibres. Water sorption was equal to or below 3.34±1.18 wt%, except for the 58 wt% fibre loading of polymer B (5.27±1.22 wt%). Water solubility was below 0.36±0.015 wt%, except for polymer B with 47 and 58 wt% fibres. For all composites, the volumetric increase was below 0.01±0.005 vol%. Residual MMA monomer was equal to or below 0.68±0.05 wt% for the fibre composites. The filter diffusion test and the (3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide) (MTT) assay demonstrated no cytotoxicity for the carbon-graphite fibre-reinforced composites, and residual cross-linking agents and vinyl chloride were not detectable by high-performance liquid chromatography (HPLC) analysis.
Sigma-Aldrich
1,4-Butanediol dimethacrylate, 95%, contains 200-300 ppm MEHQ as inhibitor