MilliporeSigma
Ultrafast irreversible phototautomerization of o-nitrobenzaldehyde.
Chemical communications (Cambridge, England) (2011-05-10)
Annapaola Migani, Verónica Leyva, Ferran Feixas, Thomas Schmierer, Peter Gilch, Inés Corral, Leticia González, Lluís Blancafort
ABSTRACT
o-Nitrobenzaldehyde is photolabile because of an irreversible phototautomerization, whereas comparable aromatic compounds function as photoprotectors because the tautomerization is reversible. In this experimental and theoretical study we track down the cause of this difference to the electronic changes that occur during the tautomerization.
Sigma-Aldrich
2-Nitrobenzaldehyde, 98%