MilliporeSigma
  • Syntheses, spectral, electrochemical and thermal studies of mononuclear manganese(III) complexes with ligands derived from 1,2-propanediamine and 2-hydroxy-3 or 5-methoxybenzaldehyde: self-assembled monolayer formation on nanostructure zinc oxide thin film.
Syntheses, spectral, electrochemical and thermal studies of mononuclear manganese(III) complexes with ligands derived from 1,2-propanediamine and 2-hydroxy-3 or 5-methoxybenzaldehyde: self-assembled monolayer formation on nanostructure zinc oxide thin film.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2011-05-04)
Mohammad Hossein Habibi, Elham Askari, Mehdi Amirnasr, Ahmad Amiri, Yuki Yamane, Takayoshi Suzuki
ABSTRACT
Mononuclear Mn(III) complexes have been prepared via the Mn(II) reaction of an equimolar of Schiff-bases derived from reaction of 2-hydroxy-3-methoxybenzaldehyde or 2-hydroxy-5-methoxybenzaldehyde with 1,2-diaminopropane. Axial ligands L include: pyridine (py) and H(2)O. The resulting complexes have been characterized by FT-IR and UV-vis spectroscopy. The crystal structures of the complexes were determined and indicate that in the solid state the complex adopts a slightly distorted octahedral environment of the imine N and hydroxo O with the two axial ligands. The electrochemical reduction of these complexes at a glassy carbon electrode in acetonitrile solution indicates that the first reduction process corresponding to Mn(III)-Mn(II) is electrochemically quasi-reversible. Thermal stability of these complexes was determined by TG and DTG. Layers of these complexes were formed on nanostructure zinc oxide thin film and a red shift was observed when zinc oxide thin film is modified by complex.
Sigma-Aldrich
(S)-(−)-1,2-Diaminopropane dihydrochloride, 99%
Sigma-Aldrich
o-Vanillin, 99%
Sigma-Aldrich
(R)-(+)-1,2-Diaminopropane dihydrochloride, 99%
Sigma-Aldrich
1,2-Diaminopropane, 99%