MilliporeSigma
  • Dimethylamine, trimethylamine, and biogenic amine formation in high-pressure processed semidried squid (Todarodes pacificius) during refrigerated storage.
Dimethylamine, trimethylamine, and biogenic amine formation in high-pressure processed semidried squid (Todarodes pacificius) during refrigerated storage.
Journal of food science (2011-05-04)
Jingyu Gou, Kun-Pyo Choi, Xinlong He, Juhee Ahn
ABSTRACT
The quality properties of semidried squid treated with high-pressure processing (HPP) were investigated during refrigerated storage. The vacuum-packed semidried squid samples were subjected to 500 MPa for 0 min (HPP-0), 5 min (HPP-5), and 10 min (HPP-10) by using a custom-made high-pressure processor and stored at a refrigerated temperature for 28 d. The populations of indigenous psychrotrophic and mesophilic bacteria were effectively reduced by approximately 1 and 2 log CFU/g at the HPP-10 treatment, respectively. Compared to the control, the bacterial counts in HPP-treated semidried squid samples were maintained at low levels throughout the storage period. The increase in the amounts of dimethylamine (DMA) and trimethylamine (TMA) was more pronounced at the unpressurized control than at the HPP treatments. The production of biogenic amines (BAs) varied with HPP treatment during refrigerated storage. Therefore, the application of HPP may provide a significant improvement in the safety and quality of semidried squid.
Sigma-Aldrich
Dimethylamine solution, purum, 33% in absolute ethanol (~5.6 M)
Sigma-Aldrich
Dimethylamine hydrochloride, purum, ≥98.0% (AT)
Sigma-Aldrich
Dimethylamine solution, 40 wt. % in H2O
Sigma-Aldrich
Dimethylamine hydrochloride, 99%
Sigma-Aldrich
Dimethylamine solution, 2.0 M in THF
Sigma-Aldrich
Dimethylamine solution, 2.0 M in methanol