MilliporeSigma
  • Influence of ground tire rubber on the transient loading response of a peat biofilter.
Influence of ground tire rubber on the transient loading response of a peat biofilter.
Journal of environmental management (2011-05-03)
F J Alvarez-Hornos, M Izquierdo, V Martínez-Soria, J M Penya-Roja, F Sempere, C Gabaldón
ABSTRACT
The effect of using ground tire rubber (GTR) as an adsorptive material in the removal of a 2:1:1 weight mixture of n-butyl acetate, toluene and m-xylene by using a peat biofilter under different intermittent conditions was investigated. The performance of two identical size biofilters, one packed with fibrous peat alone and the other with a 3:1 (vol) fibrous peat and GTR mixture, was examined for a period of four months. Partition coefficients of both materials were measured. Values of 53, 118 and 402 L kg(-1) were determined for n-butyl acetate, toluene and m-xylene in peat, respectively; and values of 40, 609 and 3035 L kg(-1) were measured for the same compounds in GTR. Intermittent load feeding of 16 h per day, 5 days per week working at an EBRT of 60 s and an inlet VOC concentration of 0.3 g C m(-1), resulted in removal efficiencies higher than 90% for both biofilters, indicating that the addition of GTR did not adversely affect the behavior of the bioreactor. Full removal of n-butyl acetate was obtained for both biofilters. GTR improved the removal of the aromatics in the first part of the biofilter, facilitating lower penetration of the toluene and m-xylene into the bed. A 31-day starvation period was applied and intermittent operation subsequently restarted. In both biofilters, high removal efficiencies after a re-acclimation period of two days were achieved. A shock loading test related to 1-h peaks of three- and four-fold increases in its baseline concentration (0.30 g C m(-3)) was applied in both biofilters. For the biofilter packed with the peat and GTR mixture, attenuation greater than 60% was observed in the maximum outlet concentration when compared to the biofilter packed with peat alone.
Sigma-Aldrich
Butyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Butyl acetate, natural, ≥98%, FG
Sigma-Aldrich
Butyl acetate, anhydrous, ≥99%
Supelco
Butyl acetate, analytical standard
Sigma-Aldrich
Butyl acetate, suitable for HPLC, 99.7%
Sigma-Aldrich
Butyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Butyl acetate, ReagentPlus®, 99.5%
Supelco
Butyl acetate, Pharmaceutical Secondary Standard; Certified Reference Material