MilliporeSigma
  • Microwave-assisted one-step synthesis of fenamic acid hydrazides from the corresponding acids.
Microwave-assisted one-step synthesis of fenamic acid hydrazides from the corresponding acids.
Molecules (Basel, Switzerland) (2011-04-30)
Tarek Aboul-Fadl, Hatem A Abdel-Aziz, Adnan Kadi, Ahmed Bari, Pervez Ahmad, Tilal Al-Samani, Seik Weng Ng
ABSTRACT
A facile and efficient method for synthesis of fenamic acid hydrazides from their acids in one-step reaction under microwave irradiation and solvent-free conditions was developed. Compared with the two-step conventional heating method, the process was simple, the reaction time was very short and the yields were almost quantitative.
Sigma-Aldrich
N-Phenylanthranilic acid, 98%