MilliporeSigma
  • Surface nanostructures in photocatalysts for visible-light-driven water splitting.
Surface nanostructures in photocatalysts for visible-light-driven water splitting.
Topics in current chemistry (2011-04-26)
Kazuhiko Maeda, Kazunari Domen
ABSTRACT
Overall water splitting to form hydrogen and oxygen over a heterogeneous (particulate) photocatalyst with solar energy is a promising process for clean and recyclable hydrogen production on a large-scale. In recent years, numerous attempts have been made for the development of photocatalysts that work under visible-light to utilize solar energy efficiently. This chapter describes recent research progress on heterogeneous photocatalysis for water splitting with visible light, particularly focusing on the development of nanostructured cocatalysts made by the authors' group.
Sigma-Aldrich
Chromium(III) oxide, nanopowder, <100 nm particle size (TEM), 98% trace metals basis
Sigma-Aldrich
Chromium(III) oxide, powder, 99.9% trace metals basis
Sigma-Aldrich
Chromium(III) oxide, powder, ≥98%