MilliporeSigma
  • Characterization of the induction and cellular role of the BaeSR two-component envelope stress response of Escherichia coli.
Characterization of the induction and cellular role of the BaeSR two-component envelope stress response of Escherichia coli.
Journal of bacteriology (2011-04-26)
Shannon K D Leblanc, Christopher W Oates, Tracy L Raivio
ABSTRACT
The bacterial cell envelope is the interface between a bacterium and its environment and is constantly exposed to environmental changes. The BaeSR two-component system regulates one of six envelope stress responses in Escherichia coli and is induced by spheroplasting, overexpression of the pilin subunit PapG, and exposure to indole. The known BaeR regulon is small, consisting of eight genes, mdtABCD-baeSR, acrD, and spy, two of which encode the BaeSR two-component system itself. In this study, we investigated the molecular nature of the BaeS-inducing cue and the cellular role of the BaeSR envelope stress response. We demonstrated that at least two flavonoids and sodium tungstate are novel inducers of the BaeSR response. Interestingly, flavonoids and sodium tungstate led to much stronger induction of the BaeSR response in an mdtA efflux pump mutant, while indole did not. These findings are consistent with the hypothesis that flavonoids and sodium tungstate are natural substrates of the MdtABC efflux pump. Indole has recently been implicated in cell-cell signaling and biofilm repression through a putative interaction with the LuxR homologue SdiA. Using genetic analyses, we found that induction of the BaeSR response by indole occurs via a pathway separate from the SdiA biofilm pathway. Further, we demonstrated that the BaeSR response does not influence biofilm formation, nor is it involved in indole-mediated inhibition of biofilm formation. We hypothesize that the main function of the Bae response is to upregulate efflux pump expression in response to specific envelope-damaging agents.
Sigma-Aldrich
Sodium tungstate dihydrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sodium tungstate dihydrate, 99.995% trace metals basis
Sigma-Aldrich
Sodium tungstate dihydrate, ACS reagent, ≥99%