MilliporeSigma
Enormous PCBs increase in oysters from the coast of Guangdong, China.
Marine pollution bulletin (2011-04-22)
Ju-Li Gan, Xiao-Ping Jia, Ting Jia, Qin Lin, Wen-Gui Cai
ABSTRACT
PCBs equivalent to Aroclor 1242 and 1254 in soft tissues of oysters Crassostrea rivularis from the Guangdong coast, China, were measured using a GC-ECD. PCB concentrations (ng g(-1)d.w.) ranged from 30 to 2040 with an average of 315 in 2003-2007, and 0.35-1.43 with an average of 0.56 in 1989-1991. Annual averages were 0.61, 0.65, 313, 290 and 342ng g(-1), respectively in 1989, 1991, 2003, 2006 and 2007. In east Guangdong, the Pearl River Estuary, and west Guangdong, regional means were 444, 273, 194ng g(-1), respectively in 2003-2007, and 0.43, 0.78, 0.38ng g(-1), respectively in 1989-1991, indicating PCB levels in oysters have risen greatly by ratios of 350-1032 compared with earlier data. PCB sources may derive from older transformers, printing materials and pressworks. Fortunately, current residual PCBs in the oysters were 0.004-0.253mg kg(-1) (w.w.), still below hygienic criteria of China and developed countries.
Product Description
Supelco
Aroclor 1242 solution, certified reference material, 50 mg/kg in transformer oil
Supelco
Aroclor 1242 solution, certified reference material, 500 mg/kg in transformer oil
Supelco
Aroclor 1254 solution, certified reference material, 500 mg/kg in transformer oil
Supelco
Aroclor 1254, ampule of 50 mg, analytical standard
Supelco
Aroclor 1254 solution, certified reference material, 50 mg/kg in transformer oil