MilliporeSigma
  • Genotoxic effects of polychlorinated biphenyls (PCB 153, 138, 101, 118) in a fish cell line (RTG-2).
Genotoxic effects of polychlorinated biphenyls (PCB 153, 138, 101, 118) in a fish cell line (RTG-2).
Toxicology in vitro : an international journal published in association with BIBRA (2011-04-21)
Laura Marabini, Rossella Calò, Serena Fucile
ABSTRACT
Polychlorinated biphenyls (PCBs) are persistent pollutants in aquatic environments, often causing the decline or disappearance of wild populations. The primary aim of this study was to investigate the genotoxic effects of some PCBs (PCB153 (2,2',4,4',5,5'-hexachlorobiphenyl) and 138 (2,2',3,4,4',5'-hexachloro-biphenyl), both non-dioxin-like compounds, and the pentachlorobiphenyls PCB118 (2,3',4,4',5-) and 101 (2,2',4',5,5'-), the former an ortho-substituted, low-affinity dioxin-like compound and the latter a non-coplanar congener classified as non-dioxin-like) in fish cells (RTG-2). These congeners are mostly present in surface waters and in edible aquatic organisms and the loss of DNA integrity in vitro serves as a sensitive biomarker of cytogenetic alterations and is considered as an initial step for the identification of genotoxic effects. The alkaline comet assay and the micronucleus test show clear genotoxic damage after short and longer exposure (2 and 24h) to maximum soluble, non-cytotoxic doses, evident sooner with PCBs 101 and 118. Oxidative stress situations involving ROS release, reduction in total GSH, lipid peroxidation and alteration to superoxide dismutase, seen after exposure with all the congeners, though with different kinetics, seem the most likely explanation for the genotoxic damage. This appears to be confirmed by the modified comet assay (pH 10) for detection of oxidized bases using endonuclease III. The increased generation of intracellular ROS might explain the apoptosis seen after treatment with the single PCBs and evaluated on the basis of the rise in 3-7 caspase activity. Therefore both the non-coplanar, non-dioxin-like PCBs (153, 138, 101) and the low-affinity dioxin-like compound PCB118 cause evident genotoxic damage, probably as a consequence of oxidative stress.
Product Description
Supelco
PCB No 101, analytical standard
2,2′,3,4,4′,5′-Hexachlorobiphenyl (IUPAC No. 138), BCR®, certified reference material
Supelco
PCB No 138, analytical standard