MilliporeSigma
  • Simultaneous determination of serine enantiomers in plasma using Mosher's reagent and stable isotope dilution gas chromatography-mass spectrometry.
Simultaneous determination of serine enantiomers in plasma using Mosher's reagent and stable isotope dilution gas chromatography-mass spectrometry.
Journal of mass spectrometry : JMS (2011-04-19)
Hiroshi Hasegawa, Yoshihiko Shinohara, Nami Masuda, Takao Hashimoto, Kimiyoshi Ichida
ABSTRACT
D-Serine is a co-agonist of the N-methyl-D-aspartate receptor in glutamate neurotransmission and has been proposed as a potential therapeutic agent for schizophrenia. However, D-serine also acts as a nephrotoxic substance in rats at high doses. To investigate the pharmacokinetics and toxicokinetics of D-serine, a method for the stereoselective determination of serine enantiomers in rat plasma was developed using GC-MS with selected ion monitoring (GC-MS-SIM). DL-[(2)H(3)]Serine was used as an internal standard to account for losses associated with the extraction, derivatization and chromatography. Serine enantiomers were purified by cation-exchange chromatography using BondElut SCX cartridge and derivatized with HCl in methanol to form methyl ester followed by subsequent N,O-diacylation with optically active (+)-α-methoxy-α-trifluoromethylphenylacetyl chloride to form epimeric amide. Quantitation was performed by SIM of the molecular-related ions of the epimers in the chemical ionization mode. The intra- and inter-day reproducibility of the assay was less than 5% for D-serine and 3% for L-serine. The method was successively applied to study the pharmacokinetics of D-serine in rats.
Sigma-Aldrich
(R)-(+)-α-Methoxy-α-trifluoromethylphenylacetic acid, 99%
Sigma-Aldrich
(S)-(−)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid, ≥99%