MilliporeSigma
Infantile haemangioma and β-blockers in otolaryngology.
European annals of otorhinolaryngology, head and neck diseases (2011-04-19)
N Leboulanger, A Cox, E-N Garabedian, F Denoyelle
ABSTRACT
Infantile haemangioma (IH) is the most common tumour during early childhood. Although these benign lesions resolve spontaneously, up until recently laryngotracheal sites of IH required invasive management. The dramatic efficacy of β-blockers on IH has radically changed the prognosis. Surgery is now no longer indicated as first-line therapy, but should only be performed for difficult, refractory cases, or in the presence of absolute contraindications to β-blockers. Long-term steroid therapy is also no longer indicated. Propranolol can be used as first-line, single-agent therapy.
Product Description
Supelco
Acebutolol hydrochloride, analytical standard
Acebutolol hydrochloride, European Pharmacopoeia (EP) Reference Standard