MilliporeSigma
  • Chiral monofluorobenzyl carbanions: synthesis of enantiopure β-fluorinated β-phenylethylamines.
Chiral monofluorobenzyl carbanions: synthesis of enantiopure β-fluorinated β-phenylethylamines.
Chemistry (Weinheim an der Bergstrasse, Germany) (2011-04-15)
José L García Ruano, Alejandro Parra, Inés Alonso, Santos Fustero, Carlos del Pozo, Yolanda Arroyo, Ascensión Sanz-Tejedor
ABSTRACT
The preparation of a stabilized monofluorobenzyl carbanion by means of a remote homochiral sulfinyl group and its completely stereoselective reactions with N-p-tolylsulfinylimines are described. The use of these reactions followed by the simultaneous removal of both chiral auxiliaries with tBuLi, which occurs without epimerization at the benzylic position, provides the quickest entry to enantiomerically pure β-fluorinated β-phenylethylamines.
Product Description
Supelco
2-Phenethylamine, analytical standard
Sigma-Aldrich
Phenethylamine, 99%
Sigma-Aldrich
Phenethylamine, ≥99%
Sigma-Aldrich
Phenethylamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
2-Phenylethylamine hydrochloride, ≥98%