MilliporeSigma
  • Effects of salt, sodium pyrophosphate, and sodium lactate on the probability of growth of Escherichia coli O157:H7 in ground beef.
Effects of salt, sodium pyrophosphate, and sodium lactate on the probability of growth of Escherichia coli O157:H7 in ground beef.
Journal of food protection (2011-04-12)
Cheng-An Hwang, Vijay Juneja
ABSTRACT
Ground beef products are susceptible to contamination with Escherichia coli O157:H7. The objective of this study was to examine the effect of salt, sodium pyrophosphate (SPP), and sodium lactate on the probability of growth of E. coli O157:H7 in ground beef under a temperature abuse condition. Ground beef containing 0 to 2.25% salt, 0 to 0.5% SPP, and 0 to 3% lactate was inoculated with a four-strain mixture of E. coli O157:H7, vacuum packaged, and stored at 10°C for 15 days. A total of 25 combinations of the three additives, each with 20 samples, were tested. A logistic regression was used to model the probability of growth of E. coli O157:H7 (with a 1.0-log CFU/g increase during storage) as a function of salt, SPP, and lactate. The resultant probability model indicated that lactate at higher concentrations decreased the probability of growth of E. coli O157:H7 in ground beef, and the effect was more pronounced at higher salt concentrations. At salt concentrations below 1.3%, the increase of SPP concentration marginally increased the growth probabilities of E. coli O157:H7. The model illustrated the effect of salt, SPP, and lactate on the growth probabilities and growth or no-growth behavior of E. coli O157:H7 in ground beef and can be used to improve the microbial food safety of ground beef products.
Sigma-Aldrich
Sodium pyrophosphate tetrabasic decahydrate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium pyrophosphate tetrabasic decahydrate, BioXtra, 99.0-103.0%
Sigma-Aldrich
Sodium pyrophosphate tetrabasic, ≥95%
Sigma-Aldrich
Sodium pyrophosphate decahydrate, ACS reagent, ≥99%
Sigma-Aldrich
Sodium pyrophosphate dibasic, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sodium DL-lactate, ReagentPlus®, ≥99.0% (NT)
Sigma-Aldrich
Sodium DL-lactate solution, syrup, 60 % (w/w), synthetic
Sigma-Aldrich
Sodium DL-lactate solution, synthetic, syrup, BioXtra, suitable for mouse embryo cell culture, 60 % (w/w)
Sigma-Aldrich
Sodium pyrophosphate dibasic, practical grade
Sigma-Aldrich
Sodium DL-lactate solution, BioReagent, syrup, 60 % (w/w), synthetic, suitable for cell culture