MilliporeSigma
  • Diasteroselective preparation of cyclopropanols using methylene bis(iodozinc).
Diasteroselective preparation of cyclopropanols using methylene bis(iodozinc).
Organic letters (2011-04-06)
Kevin Cheng, Patrick J Carroll, Patrick J Walsh
ABSTRACT
A diastereoselective synthesis of trans-2-substituted cyclopropanols is outlined. Bimetallic CH(2)(ZnI)(2) was found to react with α-chloroaldehydes to give cyclopropanols in yields of 64-89% and dr's ≥ 10:1. The high trans-selectivity resulted from equilibration of the cyclopropoxide intermediates.
Sigma-Aldrich
Diiodomethane, ReagentPlus®, 99%, contains copper as stabilizer