MilliporeSigma
  • Hydroxycamptothecin-loaded Fe3O4 nanoparticles induce human lung cancer cell apoptosis through caspase-8 pathway activation and disrupt tight junctions.
Hydroxycamptothecin-loaded Fe3O4 nanoparticles induce human lung cancer cell apoptosis through caspase-8 pathway activation and disrupt tight junctions.
Cancer science (2011-03-26)
Gen Zhang, Lei Ding, Randall Renegar, Xuemei Wang, Qun Lu, Shouquan Huo, Yan-Hua Chen
ABSTRACT
10-Hydroxycamptothecin (HCPT) elicits strong anti-cancer effects and is less toxic than camptothecin (CPT), making it widely used in recent clinical trials. However, its low solubility limits its application as an effective anti-cancer therapy. In the present study we investigate the hypothesis that the unique water dispersible oleic acid-Triton X-100-coated Fe3O4 nanoparticles loaded with HCPT disrupt epithelial cell-cell junctions and induce human lung cancer cell apoptosis through the caspase-8 pathway. We characterized the HCPT-loaded nanoparticles and determined their effects on lung cancer cell viability and apoptosis by using immunofluorescence light microscopy and SDS-PAGE/immunoblots. We found that HCPT-loaded nanoparticles elicited an anti-proliferative effect in a dose-dependent manner. HCPT-loaded nanoparticles reduced the expression of cell-cell junction protein claudins, E-cadherin and ZO-1, and transmission electron microcopy demonstrated a disrupted tight junction ultrastructure. Transepithelial electric resistance was also reduced, indicating the reduction of tight junction functions. The HCPT-loaded nanoparticles increased phosphorylation of p38 and SAPK/JNK while it showed no effects on p42/44 MAP kinase. Compared with void Fe3O4 nanoparticles or HCPT drug alone, HCPT drug-loaded nanoparticles evoked synergistic effects by increasing cell apoptosis with enhanced activation of the caspase-8 pathway. Therefore, our current study highlights the potential of HCPT drug-loaded nanoparticles as a chemotherapeutic agent for increasing anti-cancer drug efficacy.
Sigma-Aldrich
Iron(II) oxide, −10 mesh, ≥99.6% trace metals basis