MilliporeSigma
  • Silica-protected micron and sub-micron capsules and particles for self-healing at the microscale.
Silica-protected micron and sub-micron capsules and particles for self-healing at the microscale.
Macromolecular rapid communications (2011-03-25)
Aaron C Jackson, Jonathan A Bartelt, Kamil Marczewski, Nancy R Sottos, Paul V Braun
ABSTRACT
A generalized silica coating scheme is used to functionalize and protect sub-micron and micron size dicyclopentadiene monomer-filled capsules and polymer-protected Grubbs' catalyst particles. These capsules and particles are used for self-healing of microscale damage in an epoxy-based polymer. The silica layer both protects the capsules and particles, and limits their aggregation when added to an epoxy matrix, enabling the capsules and particles to be dispersed at high concentrations with little loss of reactivity.
Sigma-Aldrich
Dicyclopentadiene, contains BHT as stabilizer