MilliporeSigma
  • Fading of auramine-stained mycobacterial smears and implications for external quality assurance.
Fading of auramine-stained mycobacterial smears and implications for external quality assurance.
Journal of clinical microbiology (2011-03-25)
Jessica Minion, Shubhada Shenai, Viral Vadwai, Tejashree Tipnis, Christina Greenaway, Dick Menzies, Andrew Ramsay, Camilla Rodrigues, Madhukar Pai
ABSTRACT
Light-emitting diode fluorescence microscopy is being scaled up for tuberculosis control, but fading of auramine-stained slides could compromise external quality assurance. We stored auramine-stained slides and reexamined them over time. Slides stored in all environments faded quickly, with significant changes in the proportion of positive slides in as little as 1 week.
Sigma-Aldrich
Auramine O, Dye content 85 %, certified by the BSC
Sigma-Aldrich
Auramine O, Dye content ≥80 %, certified by the BSC