MilliporeSigma
  • HIV-negative drug addict diagnosed with AIDS and tuberculosis at autopsy: a case report and brief review of literature.
HIV-negative drug addict diagnosed with AIDS and tuberculosis at autopsy: a case report and brief review of literature.
Journal of forensic and legal medicine (2011-03-23)
Sunnassee Ananda, Zhu Shaohua, Yang Fan, Liu Liang
ABSTRACT
Human Immunodeficiency Virus (HIV) infection is a true plague and a major health concern globally. It is one of the most significant pandemics in recorded history. Despite worldwide efforts to fight the pandemic, and now with the re-emergence of tuberculosis, those clinicians, personnel performing autopsies and medical caregivers are again at risk in the work place, especially in developing countries. We describe a case where a drug abuser, whose addiction was concealed by his parents, died in hospital. He was tested HIV-negative there. A medical tangle ensued and forensic autopsy was carried out. Autopsy confirmed he was an intravenous drug addict and had tuberculosis. Post-mortem blood was positive for HIV antibodies and he was diagnosed with AIDS. Due to social stigmas, lack of knowledge or inefficient medical laboratory procedures etc, such type of cases can become a hazard to those attending the sick and to autopsy pathologists alike. We provide the case description, autopsy findings and review of pertinent literature.