MilliporeSigma
  • Direct introduction of water sample in multisegmented flow-injection analysis for sulfide determination.
Direct introduction of water sample in multisegmented flow-injection analysis for sulfide determination.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2011-03-19)
Giovana F Lima, Ariadne M Brondi, Ana L S F Paiva, César R T Tarley, André F de Oliveira, Célio Wisniewski, Pedro O Luccas
ABSTRACT
The present paper describes an inline flow-injection analysis system for the determination of sulfide in water samples, exploiting the Fischer reaction. Water samples were collected and introduced into a reactor of the FIA system. The sulfide released, after sample acidification, was carried out with a nitrogen gas flow and mixed with N,N diethyl-p-phenylenediamine (DEPD) solution in the presence of Fe(III). The blue dye formed was measured in the wavelength range between 672-679 nm. An evaluation of the effects of chemical and flow factors was performed using the factorial design of two levels, while optimization was accomplished by a Doehlert matrix. The system presented two linear-response ranges: the first of 0.433 to 400 µg L(-1) and the second of 400 to 3500 µg L(-1). The detection and quantification limit were found to be 0.130 and 0.433 µg L(-1), respectively, while the sample throughput was 12 h(-1). The precision was evaluated as the relative standard deviation (n = 10); for 50 and 100 µg L(-1) sulfide it was found to be 1.9 and 2.3%, respectively. The method showed satisfactory selectivity regarding the main interference present in environmental samples. The accuracy of the method was successfully evaluated in environmental water samples after a comparison with a literature reference method.
Sigma-Aldrich
N,N-Diethyl-p-phenylenediamine, 97%
Sigma-Aldrich
N,N-Diethyl-p-phenylenediamine sulfate salt, ≥98.0% (T)