MilliporeSigma
  • TBAF-mediated reactions of 1,1-dibromo-1-alkenes with thiols and amines and regioselective synthesis of 1,2-heterodisubstituted alkenes.
TBAF-mediated reactions of 1,1-dibromo-1-alkenes with thiols and amines and regioselective synthesis of 1,2-heterodisubstituted alkenes.
The Journal of organic chemistry (2011-03-18)
Hui Xu, Shaojin Gu, Wanzhi Chen, Dacheng Li, Jianmin Dou
ABSTRACT
An efficient synthesis of trisubstituted alkenes including 1,2-heterodisubstituted alkenes has been described. Reactions of thiols and amines with 1,1-dibromo-1-alkenes in the presence of TBAF·3H(2)O afford (Z)-2-bromovinyl sulfides and (Z)-2-bromovinyl amines regio- and stereoselectively. The reaction proceeds under catalyst-free conditions with high efficiency. The coupling reactions of the obtained products bearing bromine atoms with phenylacetylene and phenylboronic acid gave trisubstituted alkenes in good to excellent yields. Cross-coupling with various N, O, S, and P nucleophiles selectively generated 1,2-N,O, 1,2-N,S, 1,2-S,P, 1,2-S,S, and 1,2-S,O heterodisubstituted alkenes.
Sigma-Aldrich
Phenylacetylene, 98%