MilliporeSigma
  • Metal ion effect on the switch of mechanism from direct oxygen transfer to metal ion-coupled electron transfer in the sulfoxidation of thioanisoles by a non-heme iron(IV)-oxo complex.
Metal ion effect on the switch of mechanism from direct oxygen transfer to metal ion-coupled electron transfer in the sulfoxidation of thioanisoles by a non-heme iron(IV)-oxo complex.
Journal of the American Chemical Society (2011-03-18)
Jiyun Park, Yuma Morimoto, Yong-Min Lee, Wonwoo Nam, Shunichi Fukuzumi
ABSTRACT
The mechanism of sulfoxidation of thioaniosoles by a non-heme iron(IV)-oxo complex is switched from direct oxygen transfer to metal ion-coupled electron transfer by the presence of Sc(3+). The switch in the sulfoxidation mechanism is dependent on the one-electron oxidation potentials of thioanisoles. The rate of sulfoxidation is accelerated as much as 10(2)-fold by the addition of Sc(3+).
Sigma-Aldrich
Thioanisole, ReagentPlus®, ≥99%