Merck
  • Locomotor damage and brain oxidative stress induced by lead exposure are attenuated by gallic acid treatment.
Locomotor damage and brain oxidative stress induced by lead exposure are attenuated by gallic acid treatment.
Toxicology letters (2011-03-16)
Patrícia Reckziegel, Verônica Tironi Dias, Dalila Benvegnú, Nardeli Boufleur, Raquel Cristine Silva Barcelos, Hecson Jesser Segat, Camila Simonetti Pase, Clarissa Marques Moreira Dos Santos, Erico Marlon Moraes Flores, Marilise Escobar Bürger
ABSTRACT
We investigated the antioxidant potential of gallic acid (GA), a natural compound found in vegetal sources, on the motor and oxidative damages induced by lead. Rats exposed to lead (50 mg/kg, i.p., once a day, 5 days) were treated with GA (13.5mg/kg, p.o.) or EDTA (110 mg/kg, i.p.) daily, for 3 days. Lead exposure decreased the locomotor and exploratory activities, reduced blood ALA-D activity, and increased brain catalase (CAT) activity without altering other antioxidant defenses. Brain oxidative stress (OS) estimated by lipid peroxidation (TBARS) and protein carbonyl were increased by lead. GA reversed the motor behavior parameters, the ALA-D activity, as well as the markers of OS changed by lead exposure. CAT activity remained high, possibly as a compensatory mechanism to eliminate hydroperoxides during lead poisoning. EDTA, a conventional chelating agent, was not beneficial on the lead-induced motor behavior and oxidative damages. Both GA (less) and EDTA (more) reduced the lead accumulation in brain tissue. Negative correlations were observed between the behavioral parameters and lipid peroxidation and the lead levels in brain tissue. In conclusion, GA may be an adjuvant in lead exposure, mainly by its antioxidant properties against the motor and oxidative damages resulting from such poisoning.
Sigma-Aldrich
Lead(II) nitrate, 99.999% trace metals basis
Sigma-Aldrich
Lead(II) nitrate, ≥99.95% trace metals basis
Sigma-Aldrich
Lead(II) nitrate, ACS reagent, ≥99.0%