MilliporeSigma
An expedient synthesis of L-ribulose and derivatives.
Carbohydrate research (2011-03-15)
Geeta Meher, Ramanarayanan Krishnamurthy
ABSTRACT
A significant improvement in the production of l-ribulose from inexpensive and commercially available starting materials, L-arabinose and sodium aluminate, is demonstrated. This has facilitated expeditious access to gram-scale quantities of L-ribulofuranoside derivatives.
Sigma-Aldrich
L-Ribulose, ≥90% (HPLC)
Sigma-Aldrich
D-Ribulose solution, ~1 M in H2O, ≥97.0% (HPLC)