MilliporeSigma
  • Synthesis, in vitro screening and in vivo evaluation of cyclic RGD analogs cyclized through oxorhenium and oxotechnetium coordination.
Synthesis, in vitro screening and in vivo evaluation of cyclic RGD analogs cyclized through oxorhenium and oxotechnetium coordination.
European journal of medicinal chemistry (2011-03-12)
Marie Aufort, Marta Gonera, Marie-Anne Lelait, Bertrand Czarny, Loïc Le Clainche, Robert Thaï, Amandine Landra, Mathias Ruinart de Brimont, Christophe Dugave
ABSTRACT
A library of RGD tripeptide analogs cyclized through oxorhenium coordination by an NS2/S chelation motif was synthesized. Screening towards integrins αVβ3, αIIbβ3 and αVβ5 led to the identification of 6 oxorhenium complexes that bind to integrin αVβ3 in the submicromolar range. In vivo evaluation of five of the corresponding oxotechnetium complexes using nude mice bearing a U87MG human tumor xenograft showed a significant and specific accumulation of radioactivity inside the tumor. The best results in vivo were obtained with complexes Tc-16 and Tc-50 that displayed a higher tumor accumulation and a lower distribution in other tissues relative to a reference cyclopentapeptide tracer.
Sigma-Aldrich
Potassium perrhenate, 99.98% trace metals basis
Sigma-Aldrich
Potassium perrhenate, 99%
Sigma-Aldrich
Ammonium perrhenate, ≥99%
Sigma-Aldrich
Ammonium perrhenate, 99.999% trace metals basis
Sigma-Aldrich
Sodium perrhenate, 99.99% trace metals basis