MilliporeSigma
  • N-nitro-L-arginine, a nitric oxide synthase inhibitor, aggravates iminodipropionitrile-induced neurobehavioral and vestibular toxicities in rats.
N-nitro-L-arginine, a nitric oxide synthase inhibitor, aggravates iminodipropionitrile-induced neurobehavioral and vestibular toxicities in rats.
Experimental and toxicologic pathology : official journal of the Gesellschaft fur Toxikologische Pathologie (2011-03-11)
Haseeb Ahmad Khan
ABSTRACT
Exposure of iminodipropionitrile (IDPN) to rodents produces permanent behavioral syndrome characterized by repetitive head movements, circling and back walking. Other synthetic nitriles of industrial importance such as crotonitrile and allylnitrile are also able to produce similar motor deficits in experimental animals. However, due to the well-defined behavioral deficits and their easy quantification, IDPN-induced behavioral syndrome is a preferential animal model to test the interaction of various agents with synthetic nitriles. This study reports the effect of non-specific nitric oxide synthase inhibitor, N-nitro-L-arginine (NARG) on IDPN-induced neurobehavioral toxicity in adult male Wistar rats. Four groups of animals were given i.p. injections of IDPN (100 mg/kg) for 6 days. These rats were treated with oral administration of NARG in the doses of 0 (IDPN alone group), 50, 150 and 300 mg/kg, 60 min before IDPN, respectively. Control rats received vehicle only, whereas another group was treated with 300 mg/kg of NARG alone (without IDPN). The results showed that NARG significantly exacerbated the incidence and intensity of IDPN-induced dyskinetic head movements, circling and back walking. The histology of inner ear showed massive degeneration of the sensory hair cells in the crista ampullaris of rats receiving the combined treatment with IDPN and NARG, suggesting a possible role of nitric oxide in IDPN-induced neurobehavioral syndrome in rats.
Sigma-Aldrich
3,3′-Iminodipropionitrile, technical grade, 90%