MilliporeSigma
  • ATP selective acridone based fluorescent probes for monitoring of metabolic events.
ATP selective acridone based fluorescent probes for monitoring of metabolic events.
Chemical communications (Cambridge, England) (2011-03-08)
Jatinder Kaur, Palwinder Singh
ABSTRACT
Acridones carrying an appropriate substituent at N-10 showed significant fluorescence changes on interacting with ATP in HEPES buffer at pH 7.2. The selectivity and sufficient binding of these probes with ATP could be useful for monitoring of metabolic processes.
Sigma-Aldrich
9(10H)-Acridanone, 99%