MilliporeSigma
  • First prebiotic generation of a ribonucleotide from adenine, D-ribose and trimetaphosphate.
First prebiotic generation of a ribonucleotide from adenine, D-ribose and trimetaphosphate.
Chemical communications (Cambridge, England) (2011-02-10)
Graziano Baccolini, Carla Boga, Gabriele Micheletti
ABSTRACT
Adenosine monophosphate isomers are obtained by self-assembling of adenine, D-ribose and trimetaphosphate in aqueous solution in good yields. This generation of a ribonucleotide from its three molecular components occurs in a one-pot reaction at room temperature for about 30-40 days and with high chemio-, regio-, and stereo-selectivity. Similar results are obtained with guanine. A mechanism is also proposed.
Sigma-Aldrich
Trisodium trimetaphosphate, ≥95%