MilliporeSigma
  • A new and rapid method for the assay of bacterial fatty acids using high resolution capillary gas chromatography and trimethylsulfonium hydroxide.
A new and rapid method for the assay of bacterial fatty acids using high resolution capillary gas chromatography and trimethylsulfonium hydroxide.
Zentralblatt fur Bakteriologie : international journal of medical microbiology (1990-11-01)
K D Müller, H Husmann, H P Nalik
ABSTRACT
Gas-liquid chromatography of cellular fatty acids is a useful tool for the identification of bacteria. Derivatization of bacterial fatty acids to methyl esters by conventional techniques is usually time-consuming and complicated. A new one-step technique using trimethyl-sulfonium hydroxide allows the direct formation of fatty acid methyl esters within 1-2 min. Some random examples of profiles demonstrate that straight, branched, saturated, unsaturated, hydroxy and cyclopropyl fatty acids match conventional preparations well. The method is a very sensitive one, since only a few colonies are sufficient for preparation of fatty acid methyl esters.
Sigma-Aldrich
Trimethylsulfonium iodide, 98%