MilliporeSigma
Thermal and electron-induced decomposition of 2-butanol on Pt(111).
The journal of physical chemistry. A (2011-01-26)
Xiaofeng Hu, Richard A Rosenberg, Michael Trenary
ABSTRACT
The adsorption, thermal evolution, and electron irradiation of 2-butanol on Pt(111) were investigated with reflection absorption infrared spectroscopy (RAIRS). A simulated vibrational spectrum of a single 2-butanol molecule was calculated using density functional theory to facilitate vibrational assignments. Exposures of 0.2 Langmuir (L) and lower result in both isolated 2-butanol molecules with minimal lateral interactions and hydrogen-bonded clusters. The thermal evolution following a 4.0 L exposure shows that the hydrogen-bonded multilayer desorbs around 170 K, leaving a 2-butanol monolayer where hydrogen bonding still exists. At 190 K, a new feature at 1699 cm(-1) is attributed to the formation of butanone. Irradiation with 750 or 100 eV electrons leads to 2-butanol desorption and partial conversion to butanone, as indicated by the appearance of a peak at 1709 cm(-1).
Sigma-Aldrich
(S)-(+)-2-Butanol, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Aluminum-tri-sec-butoxide, 97%
Pricing and availability is not currently available.
Supelco
2-Butanol, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
2-Butanol, anhydrous, 99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
2-Butanol, ReagentPlus®, ≥99%
Pricing and availability is not currently available.