MilliporeSigma
  • Palladium catalyzed bicyclization of 1,8-diiodonaphthalene and tertiary propargylic alcohols to phenalenones and their applications as fluorescent chemosensor for fluoride ions.
Palladium catalyzed bicyclization of 1,8-diiodonaphthalene and tertiary propargylic alcohols to phenalenones and their applications as fluorescent chemosensor for fluoride ions.
Chemical communications (Cambridge, England) (2011-01-15)
Xiaopeng Chen, Hongbo Wang, Xiaohan Jin, Jinwu Feng, Yanguang Wang, Ping Lu
ABSTRACT
Phenalenone derivatives were efficiently constructed from 1,8-diiodonaphthalene and tertiary propynols via a one-pot domino reaction which eventually included Pd-catalyzed Sonogoshira coupling, Pd-catalyzed allylic oxidation and Pd-catalyzed C(sp(2))-H activation. Moreover, the synthesized phenalenone derivative presented a practical application as a fluorescent chemosensor for fluoride anion with high sensitivity and selectivity.
Sigma-Aldrich
Perinaphthenone, 97%