MilliporeSigma
  • 2,3-Dimethyl-2-butene (TME) ozonolysis: pressure dependence of stabilized Criegee intermediates and evidence of stabilized vinyl hydroperoxides.
2,3-Dimethyl-2-butene (TME) ozonolysis: pressure dependence of stabilized Criegee intermediates and evidence of stabilized vinyl hydroperoxides.
The journal of physical chemistry. A (2010-12-18)
Greg T Drozd, Jesse Kroll, Neil M Donahue
ABSTRACT
We present measurements of the pressure dependence of stabilized Criegee intermediate (SCI) formation utilizing a hexafluoroacetone scavenger. SCI yields in the ozonolysis of 2,3-dimethyl-2-butene (TME) were measured in a high pressure flow reactor within a range of 50-710 Torr. Within this pressure range, SCI yields increase linearly with pressure. A zero pressure intercept of about 15% indicates that a significant fraction of CI are formed below the barrier to isomerization. By comparison of our results of the pressure dependence of SCI formation and both prompt and long-time OH yields, our results indicate that OH formation from ozonolysis proceeds via at least two intermediates, the SCI and presumably a vinylhydroperoxide (VHP).
Sigma-Aldrich
Hexafluoroacetone trihydrate, 98%
Sigma-Aldrich
2,3-Dimethyl-2-butene, ≥99%
Sigma-Aldrich
2,3-Dimethyl-2-butene, 98%