MilliporeSigma
  • Large-scale production of magnetosomes by chemostat culture of Magnetospirillum gryphiswaldense at high cell density.
Large-scale production of magnetosomes by chemostat culture of Magnetospirillum gryphiswaldense at high cell density.
Microbial cell factories (2010-12-15)
Yang Liu, Guo R Li, Fang F Guo, Wei Jiang, Ying Li, Lun J Li
ABSTRACT
Magnetotactic bacteria have long intrigued researchers because they synthesize intracellular nano-scale (40-100 nm) magnetic particles composed of Fe3O4, termed magnetosomes. Current research focuses on the molecular mechanisms of bacterial magnetosome formation and its practical applications in biotechnology and medicine. Practical applications of magnetosomes are based on their ferrimagnetism, nanoscale size, narrow size distribution, dispersal ability, and membrane-bound structure. However, the applications of magnetosomes have not yet been developed commercially, mainly because magnetotactic bacteria are difficult to cultivate and consistent, high yields of magnetosomes have not yet been achieved. We report a chemostat culture technique based on pH-stat feeding that yields a high cell density of Magnetospirillum gryphiswaldense strain MSR-1 in an auto-fermentor. In a large-scale fermentor, the magnetosome yield was significantly increased by adjusting the stirring rate and airflow which regulates the level of dissolved oxygen (DO). Low concentration of sodium lactate (2.3 mmol l-1) in the culture medium resulted in more rapid cell growth and higher magnetosome yield than high concentration of lactate (20 mmol l-1). The optical density of M. gryphiswaldense cells reached 12 OD565 nm after 36 hr culture in a 42 L fermentor. Magnetosome yield and productivity were 83.23 ± 5.36 mg l-1 (dry weight) and 55.49 mg l-1 day-1, respectively, which were 1.99 and 3.32 times higher than the corresponding values in our previous study. Compared to previously reported methods, our culture technique with the MSR-1 strain significantly increased cell density, cell yield, and magnetosome yield in a shorter time window and thus reduced the cost of production. The cell density and magnetosome yield reported here are the highest so far achieved with a magnetotactic bacteria. Refinement of this technique will enable further increase of cell density and magnetosome yield.
Sigma-Aldrich
Sodium DL-lactate, ReagentPlus®, ≥99.0% (NT)
Sigma-Aldrich
Sodium DL-lactate solution, syrup, 60 % (w/w), synthetic
Sigma-Aldrich
Sodium DL-lactate solution, synthetic, syrup, BioXtra, suitable for mouse embryo cell culture, 60 % (w/w)
Sigma-Aldrich
Sodium DL-lactate solution, BioReagent, syrup, 60 % (w/w), synthetic, suitable for cell culture