MilliporeSigma
  • Enantioselective Morita-Baylis-Hillman reaction of isatins with acrylates: facile creation of 3-hydroxy-2-oxindoles.
Enantioselective Morita-Baylis-Hillman reaction of isatins with acrylates: facile creation of 3-hydroxy-2-oxindoles.
Organic letters (2010-12-09)
Fangrui Zhong, Guo-Ying Chen, Yixin Lu
ABSTRACT
The first tertiary amine catalyzed enantioselective Morita-Baylis-Hillman (MBH) reaction of isatins with acrylates has been demonstrated, allowing asymmetric synthesis of biologically significant 3-substituted-3-hydroxy-2-oxindoles in good yields and with excellent enantioselectivities. The C6'-OH group of β-isocupreidine (β-ICD) is believed to facilitate the key proton transfer step in the MBH reaction, via an intramolecular proton relay process.
Sigma-Aldrich
2-Oxindole, 97%
Pricing and availability is not currently available.