MilliporeSigma
  • In vitro toxicity of bithionol and bithionol sulphoxide to Neoparamoeba spp., the causative agent of amoebic gill disease (AGD).
In vitro toxicity of bithionol and bithionol sulphoxide to Neoparamoeba spp., the causative agent of amoebic gill disease (AGD).
Diseases of aquatic organisms (2010-12-08)
Renee L Florent, Joy A Becker, Mark D Powell
ABSTRACT
The objective of the present study was to evaluate the in vitro toxicity of bithionol and bithionol sulphoxide to Neoparamoeba spp., the causative agent of amoebic gill disease (AGD). The current treatment for AGD-affected Atlantic salmon involves bathing sea-caged fish in freshwater for a minimum of 3 h, a labour-intensive and costly exercise. Previous attempts to identify alternative treatments have suggested bithionol as an alternate therapeutic, but extensive in vitro efficacy testing has not yet been done. In vitro toxicity to Neoparamoeba spp. was examined using amoebae isolated from the gill of AGD-affected Atlantic salmon and exposing the parasites to freshwater, alumina (10 mg l(-1)), seawater, bithionol or bithionol sulphoxide at nominal concentrations of 0.1, 0.5, 1, 5 and 10 mg l(-1) in seawater. The numbers of viable amoebae were counted using the trypan blue exclusion method at 0, 24, 48 and 72 h. Both bithionol and bithionol sulphoxide demonstrated in vitro toxicity to Neoparamoeba spp. at all concentrations examined (0.1 to 10 mg l(-1) over 72 h), with a comparable toxicity to freshwater observed for both chemicals at concentrations > 5 mg l(-1) following a 72 h treatment. Freshwater remained the most effective treatment, with only 6% viable amoebae seen after 24 h and no viable amoebae observed after 48 h.
Product Description
Supelco
Bithionol, VETRANAL®, analytical standard