MilliporeSigma
  • Characterization of multiple chlorobenzoic acid-degrading organisms from pristine and contaminated systems: mineralization of 2,4-dichlorobenzoic acid.
Characterization of multiple chlorobenzoic acid-degrading organisms from pristine and contaminated systems: mineralization of 2,4-dichlorobenzoic acid.
Bioresource technology (2010-11-16)
Sunday A Adebusoye, Marzia Miletto
ABSTRACT
Multiple bacterial strains with CBA metabolic properties were isolated using a simple selective strategy. Phylogenetic analysis of the 16S rRNA gene sequences grouped them into two main clusters consisting of four bacterial phyla and belonging to 17 genera. Whereas growth was more frequent with 2-CBA (∼68%), 50% grew on 4-CBA and ∼7% utilized 3-CBA. One third of the strains exhibited 2,4-dichlorobenzoic acid (2,4-diCBA) catabolic function and were mainly representatives of α-, β- and γ-Proteobacteria. In batch experiments, growth was concomitant with substrate disappearance and near-stoichiometric release of chloride. Doubling times for 2,4-diCBA degradation doubled those determined for mono-substituted CBAs. Out of the six 2,4-diCBA degraders submitted for enzyme assays, significant induction of catechol 1,2-dioxygenase types I and II activities in cell-free extracts were found in four while protocatechuate 3,4-dioxygenase activity was detected in the remaining two. Activities in CBA-grown cells were 20 orders-of-magnitude higher than those grown on benzoic acid.
Product Description
Supelco
2,4-Dichlorobenzoic acid, PESTANAL®, analytical standard
Sigma-Aldrich
2,4-Dichlorobenzoic acid, 98%