MilliporeSigma
  • Composition and intraspecific chemical variability of the essential oil from Artemisia herba-alba growing wild in a Tunisian arid zone.
Composition and intraspecific chemical variability of the essential oil from Artemisia herba-alba growing wild in a Tunisian arid zone.
Chemistry & biodiversity (2010-11-13)
Hédi Mighri, Ahmed Akrout, Hajer El-jeni, Slah Zaidi, Félix Tomi, Joseph Casanova, Mohamed Neffati
ABSTRACT
The intraspecific chemical variability of essential oils (50 samples) isolated from the aerial parts of Artemisia herba-alba Asso growing wild in the arid zone of Southeastern Tunisia was investigated. Analysis by GC (RI) and GC/MS allowed the identification of 54 essential oil components. The main compounds were β-thujone and α-thujone, followed by 1,8-cineole, camphor, chrysanthenone, trans-sabinyl acetate, trans-pinocarveol, and borneol. Chemometric analysis (k-means clustering and PCA) led to the partitioning into three groups. The composition of two thirds of the samples was dominated by α-thujone or β-thujone. Therefore, it could be expected that wild plants of A. herba-alba randomly harvested in the area of Kirchaou and transplanted by local farmers for the cultivation in arid zones of Southern Tunisia produce an essential oil belonging to the α-thujone/β-thujone chemotype and containing also 1,8-cineole, camphor, and trans-sabinyl acetate at appreciable amounts.
Sigma-Aldrich
(−)-α-Thujone, ≥96.0% (GC)