MilliporeSigma
  • Activation of brain histaminergic neurotransmission: a mechanism for cognitive effects of memantine in Alzheimer's disease.
Activation of brain histaminergic neurotransmission: a mechanism for cognitive effects of memantine in Alzheimer's disease.
The Journal of pharmacology and experimental therapeutics (2010-11-09)
M Motawaj, A Burban, E Davenas, J-M Arrang
ABSTRACT
We previously reported that some N-methyl-D-aspartate (NMDA)-receptor antagonists enhanced histamine neuron activity in rodents. Here, we have investigated the effects of memantine, an NMDA-receptor antagonist used for the treatment of Alzheimer's disease, on histaminergic neurotransmission. In vitro, memantine antagonized native NMDA receptors with a micromolar potency but had no effect at recombinant human histamine receptors. In vivo, a single administration of memantine increased histamine neuron activity, as shown by the 60% increase of tele-methylhistamine (t-MeHA) levels observed in the brain of mice. This increase occurred with an ED(50) of 0.3 ± 0.1 mg/kg, similar to that found on inhibition of ex vivo [(3)H]dizocilpine maleate (MK-801) binding (1.8 ± 1.3 mg/kg). Two days after pretreatment of mice with memantine at 5 mg/kg twice daily for 5 days, t-MeHA levels were enhanced by 50 ± 7% (p < 0.001), indicating a long-lasting activation of histamine neurons. Quantitative polymerase chain reaction analysis was used to explore genes involved in this persistent effect. H(3) receptor mRNAs were strongly increased, but the density of H(3) receptor binding sites was increased solely in hypothalamus (by 141 ± 24%). Up-regulations of brain-derived neurotrophic factor and NMDA-receptor 1 subunit mRNAs were also found but were restricted to hippocampus. mRNA expression of α7-nicotinic receptors remained unchanged in any region. Considering the well established cognitive effects of histamine neurons, the increase in brain t-MeHA levels after single or repeated administration of therapeutic doses of memantine suggests that the drug exerts its beneficial effects on cognitive deficits of Alzheimer's disease, at least partly, by activating histamine neurons.
Sigma-Aldrich
1-Methylhistamine dihydrochloride, ≥98% (TLC), powder