MilliporeSigma
  • Synthesis of oligoribonucleic acid conjugates using a cyclooctyne phosphoramidite.
Synthesis of oligoribonucleic acid conjugates using a cyclooctyne phosphoramidite.
Organic letters (2010-11-06)
Pieter van Delft, Nico J Meeuwenoord, Sascha Hoogendoorn, Jasper Dinkelaar, Herman S Overkleeft, Gijsbert A van der Marel, Dmitri V Filippov
ABSTRACT
The conjugation of a ribonucleic acid 16-mer with the cationic amphiphilic peptide penetratin and an anionic hyaluronan tetrasaccharide by means of Cu-free "click" chemistry is reported. The alkyne-functionalized 16-mer was prepared by automated solid-phase synthesis, using a newly developed strained cyclooctyne phosphoramidite in the final coupling. Cycloaddition of the alkyne functionalized RNA to the azide containing biomolecules led to a clean conversion into the corresponding nucleic acid conjugates.
Sigma-Aldrich
Cyclooctane, ≥99%