MilliporeSigma
  • Preparation and application of rifamycin-capped (3-(2-O-β-cyclodextrin)-2-hydroxypropoxy)-propylsilyl-appended silica particles as chiral stationary phase for high-performance liquid chromatography.
Preparation and application of rifamycin-capped (3-(2-O-β-cyclodextrin)-2-hydroxypropoxy)-propylsilyl-appended silica particles as chiral stationary phase for high-performance liquid chromatography.
Talanta (2010-11-03)
Jia Zhao, Daphane Tan, S K Thamarai Chelvi, E L Yong, Hian Kee Lee, Yinhan Gong
ABSTRACT
Rifamycin-capped (3-(2-O-β-cyclodextrin)-2-hydroxypropoxy)-propylsilyl-appended silica particles (RCD-HPS), a new type of substituted β-cyclodextrin-bonded chiral stationary phase (CSP) for high-performance liquid chromatography (HPLC), have been synthesized by the treatment of bromoacetate-substituted-(3-(2-O-β-cyclodextrin)-2-hydroxypropoxy)-propylsilyl-appended silica particles (BACD-HPS) with rifamycin SV in anhydrous acetonitrile. The stationary phase is characterized by means of elemental analysis and Fourier-transform infrared spectroscopy. This new CSP has a chiral selector with two recognition sites: rifamycin and β-cyclodextrin (β-CD). The chromatographic behavior of RCD-HPS was studied with several disubstituted benzenes and some chiral drug compounds under reversed-phase HPLC mobile phase conditions. The results show that RCD-HPS has excellent selectivity for the separation of aromatic positional isomers and enantiomers of chiral compounds due to the cooperative functioning of rifamycin and β-CD.
Product Description
Rifamycin sodium, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Rifamycin SV sodium salt, potency: ≥ 900 IU/mg (anhydrous basis)