MilliporeSigma
  • Synthesis of novel α-santonin derivatives as potential cytotoxic agents.
Synthesis of novel α-santonin derivatives as potential cytotoxic agents.
European journal of medicinal chemistry (2010-10-26)
Francisco F P Arantes, Luiz C A Barbosa, Célia R A Maltha, Antônio J Demuner, Patricia Marçal da Costa, José R O Ferreira, Letícia V Costa-Lotufo, Manoel O Moraes, Cláudia Pessoa
ABSTRACT
Ten novel α-santonin derivatives have been synthesized as cytotoxic agents. The in vitro antitumor activity of these compounds has been evaluated against cancer cells lines. Structure-activity relationships indicate that α-methylene-γ-lactone and endoperoxide functionalities play important roles in conferring cytotoxicity. The compounds 2-4, possessing the α-methylene-γ-lactone group showed IC50 values between 5.70 and 16.40 μM. Mixture of isomers 5 and 6, with the α-methylene-γ-lactone and endoperoxide functionalities, displayed the greatest activity, with IC50 values between 1.45 and 4.35 μM. The biological assays conducted with normal cells revealed that the compounds 2, 5 and 6 are selective against cancer cells lines tested. Bioactive lactones described herein and in our previous report did not cause disruption of the cell membrane in mouse erythrocytes.