MilliporeSigma
  • Diastereoselective nitrenium ion-mediated cyclofunctionalization: total synthesis of (+)-castanospermine.
Diastereoselective nitrenium ion-mediated cyclofunctionalization: total synthesis of (+)-castanospermine.
Organic letters (2010-10-23)
Edward G Bowen, Duncan J Wardrop
ABSTRACT
The asymmetric total synthesis of the α-glucosidase inhibitor (+)-castanospermine is reported. The central theme in our approach to this polyhydroxylated alkaloid is the simultaneous generation of the piperidine ring and the C-1/8a erythro stereodiad through the diastereoselective, oxamidation of an unsaturated O-alkyl hydroxamate. This process is believed to proceed sequentially via singlet acylnitrenium and aziridinium ion intermediates.
Sigma-Aldrich
(1S,6S,7R,8R,8aR)-1,6,7,8-Tetrahydroxyindolizidine, 98%