MilliporeSigma
  • The role of serotonin in the enhancement of long-term memory resulting from predator detection in Lymnaea.
The role of serotonin in the enhancement of long-term memory resulting from predator detection in Lymnaea.
The Journal of experimental biology (2010-10-19)
Jae Il-Han, Tara Janes, Ken Lukowiak
ABSTRACT
Serotonergic systems play important roles in modulating stress-induced arousal and vigilance behaviours. The pond snail, Lymnaea, shows multiple defensive vigilance behaviours in response to the stress associated with predator detection. Predator detection elicited by crayfish effluent (CE), increases the time to re-emerge from the shell and enhances the shadow withdrawal response. More importantly, in Lymnaea, CE enhances the ability to form long-term memory (LTM). We investigated the role of the serotonergic system in these anti-predator responses in Lymnaea. Using a serotonin-receptor antagonist, mianserin, we found that two defensive vigilance behaviours (e.g. increasing the time to re-emerge from their shell and shadow response) elicited by CE were not observed when the serotonergic system was disrupted. Also, methysergide, another serotonin antagonist, blocked the enhanced LTM formation after training in CE. Importantly, mianserin did not alter LTM formation in pond water (PW). These data suggest that a serotonergic system is activated only when Lymnaea detect a predator. When snails were trained in CE using a training procedure that in PW produces a 24-h LTM, a more persistent form of LTM (5 days) occurred. This more persistent form of LTM was abolished after mianserin treatment. Increasing 5-HT levels in the snail by the injection of 5-HT was also associated with enhanced LTM formation. Lastly, we tested whether the osphradium is implicated in CE detection and subsequent enhanced formation of LTM. Cutting the osphradial nerve to the CNS resulted in the loss of the ability to form enhanced LTM in CE. Together, these findings support the hypothesis that the serotonergic system plays a key role in modulating the predator-induced stress responses in Lymnaea.
Sigma-Aldrich
Methysergide maleate salt, solid