MilliporeSigma
  • Influence of concentration and temperature on the dynamics of water in the hydrophobic hydration shell of tetramethylurea.
Influence of concentration and temperature on the dynamics of water in the hydrophobic hydration shell of tetramethylurea.
Journal of the American Chemical Society (2010-10-19)
Klaas-Jan Tielrooij, Johannes Hunger, Richard Buchner, Mischa Bonn, Huib J Bakker
ABSTRACT
We study the influence of the amphipilic compound tetramethylurea (TMU) on the dynamical properties of water, using dielectric relaxation spectroscopy in the regime between 0.2 GHz and 2 THz. This technique is capable of resolving different water species, their relative fractions, and their corresponding reorientation dynamics. We find that the reorientation dynamics of water molecules in the hydration shell of the hydrophobic groups of TMU is between 3 (at low concentrations) and 10 (at higher concentrations) times slower than the dynamics of bulk water. The data indicate that the effect of hydrophobic groups on water is strong but relatively short-ranged. With increasing temperature, the fraction of water contained in the hydrophobic hydration shell decreases, which implies that the overall effect of hydrophobic groups on water becomes smaller.
Sigma-Aldrich
Tetramethylurea, 99%